RIE240871008 Rieke Metals, Inc.

2-Chloro-5-(3,5-difluorobenzoyl)pyridine

  • MDL number MFCD07775842
  • Molecular Weight : 253.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5625-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-42
Availability details for
Quantity
  • Availability
  • Add to Cart