RIE240871014 Rieke Metals, Inc.

2-Chloro-5-(3,5-dichlorobenzoyl)pyridine

  • MDL number MFCD07775840
  • Molecular Weight : 286.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-chloro-3-pyridinyl)(3,5-dichlorophenyl)methanone
Inchi Code 1S/C12H6Cl3NO/c13-9-3-8(4-10(14)5-9)12(17)7-1-2-11(15)16-6-7/h1-6H
Inchi Key FZYKWODROKLMON-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5625-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-48
Availability details for
Quantity
  • Availability
  • Add to Cart