RIE240871015 Rieke Metals, Inc.

2-Chloro-5-(2,3-dimethoxybenzoyl)pyridine

  • MDL number MFCD13153380
  • Molecular Weight : 277.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5625-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-49
Availability details for
Quantity
  • Availability
  • Add to Cart