RIE240871023 Rieke Metals, Inc.

2-Chloro-5-(2,5-dimethylbenzoyl)pyridine

  • MDL number MFCD13153387
  • Molecular Weight : 245.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-chloro-3-pyridinyl)(2,5-dimethylphenyl)methanone
Inchi Code 1S/C14H12ClNO/c1-9-3-4-10(2)12(7-9)14(17)11-5-6-13(15)16-8-11/h3-8H,1-2H3
Inchi Key YDTUTMDUOIGWOB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5625-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-57
Availability details for
Quantity
  • Availability
  • Add to Cart