RIE240871032 Rieke Metals, Inc.

3-(3-bromophenyl)-2'-methoxypropiophenone

  • MDL number MFCD03843301
  • Molecular Weight : 319.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1800-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1800-05
Availability details for
Quantity
  • Availability
  • Add to Cart