RIE240871033 Rieke Metals, Inc.

3-(3-bromophenyl)-3'-methoxypropiophenone

  • MDL number MFCD03843302
  • Molecular Weight : 319.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-bromophenyl)-1-(3-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H15BrO2/c1-19-15-7-3-5-13(11-15)16(18)9-8-12-4-2-6-14(17)10-12/h2-7,10-11H,8-9H2,1H3
Inchi Key WVHXPWAUNKPSFQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1800-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1800-06
Availability details for
Quantity
  • Availability
  • Add to Cart