RIE240871037 Rieke Metals, Inc.

3-(3-bromophenyl)-4'-cyanopropiophenone

  • MDL number MFCD03843306
  • Molecular Weight : 314.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[3-(3-bromophenyl)propanoyl]benzonitrile
Inchi Code 1S/C16H12BrNO/c17-15-3-1-2-12(10-15)6-9-16(19)14-7-4-13(11-18)5-8-14/h1-5,7-8,10H,6,9H2
Inchi Key XZTIYPZOWJPDEF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1800-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1800-10
Availability details for
Quantity
  • Availability
  • Add to Cart