RIE240871040 Rieke Metals, Inc.

3-(3-bromophenyl)-4'-carboethoxypropiophenone

  • MDL number MFCD03843309
  • Molecular Weight : 361.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1800-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1800-13
Availability details for
Quantity
  • Availability
  • Add to Cart