RIE240871044 Rieke Metals, Inc.

3-(3-bromophenyl)-3'-chloropropiophenone

  • MDL number MFCD03843314
  • Molecular Weight : 323.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1800-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1800-20
Availability details for
Quantity
  • Availability
  • Add to Cart