RIE240871045 Rieke Metals, Inc.

3-(3-bromophenyl)-4'-chloropropiophenone

  • MDL number MFCD03843315
  • Molecular Weight : 323.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1800-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1800-21
Availability details for
Quantity
  • Availability
  • Add to Cart