RIE240871048 Rieke Metals, Inc.

3-(3-bromophenyl)-2',3'-dimethylpropiophenone

  • MDL number MFCD03843318
  • Molecular Weight : 317.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-bromophenyl)-1-(2,3-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H17BrO/c1-12-5-3-8-16(13(12)2)17(19)10-9-14-6-4-7-15(18)11-14/h3-8,11H,9-10H2,1-2H3
Inchi Key ZMWPLGFJPCKMJX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1800-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1800-26
Availability details for
Quantity
  • Availability
  • Add to Cart