RIE240871051 Rieke Metals, Inc.

3-(3-bromophenyl)-2',6'-dimethylpropiophenone

  • MDL number MFCD03843321
  • Molecular Weight : 317.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1800-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1800-29
Availability details for
Quantity
  • Availability
  • Add to Cart