RIE240871053 Rieke Metals, Inc.

3-(3-bromophenyl)-3',5'-dimethylpropiophenone

  • MDL number MFCD03843323
  • Molecular Weight : 317.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-bromophenyl)-1-(3,5-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H17BrO/c1-12-8-13(2)10-15(9-12)17(19)7-6-14-4-3-5-16(18)11-14/h3-5,8-11H,6-7H2,1-2H3
Inchi Key GYCGDXVBYJCOGC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1800-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1800-31
Availability details for
Quantity
  • Availability
  • Add to Cart