RIE240871054 Rieke Metals, Inc.

4'-bromo-3-(3-bromophenyl)-3'-fluoropropiophenone

  • MDL number MFCD03843324
  • Molecular Weight : 386.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1800-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1800-32
Availability details for
Quantity
  • Availability
  • Add to Cart