RIE240871057 Rieke Metals, Inc.

3-(3-bromophenyl)-2'-chloropropiophenone

  • MDL number MFCD03843327
  • Molecular Weight : 323.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-bromophenyl)-1-(2-chlorophenyl)-1-propanone
Inchi Code 1S/C15H12BrClO/c16-12-5-3-4-11(10-12)8-9-15(18)13-6-1-2-7-14(13)17/h1-7,10H,8-9H2
Inchi Key GZKFPUNUVMVVRA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1800-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1800-36
Availability details for
Quantity
  • Availability
  • Add to Cart