RIE240871062 Rieke Metals, Inc.

4'-bromo-3-(3-bromophenyl)-2'-fluoropropiophenone

  • MDL number MFCD03843332
  • Molecular Weight : 386.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1800-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1800-49
Availability details for
Quantity
  • Availability
  • Add to Cart