RIE240871074 Rieke Metals, Inc.

3-(3-bromophenyl)-3',4',5'-trifluoropropiophenone

  • MDL number MFCD03843344
  • Molecular Weight : 343.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-bromophenyl)-1-(3,4,5-trifluorophenyl)-1-propanone
Inchi Code 1S/C15H10BrF3O/c16-11-3-1-2-9(6-11)4-5-14(20)10-7-12(17)15(19)13(18)8-10/h1-3,6-8H,4-5H2
Inchi Key KOACLFHPFYUJBO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1800-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1800-61
Availability details for
Quantity
  • Availability
  • Add to Cart