RIE240871075 Rieke Metals, Inc.

3-(3-Bromophenyl)-2',6'-dichloropropiophenone

  • MDL number MFCD08064136
  • Molecular Weight : 358.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-bromophenyl)-1-(2,6-dichlorophenyl)-1-propanone
Inchi Code 1S/C15H11BrCl2O/c16-11-4-1-3-10(9-11)7-8-14(19)15-12(17)5-2-6-13(15)18/h1-6,9H,7-8H2
Inchi Key PUIPEGPHPRKXQH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1800-62

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1800-62
Availability details for
Quantity
  • Availability
  • Add to Cart