RIE240871078 Rieke Metals, Inc.

2-(3-bromophenyl)ethyl cyclopentyl ketone

  • MDL number MFCD03843347
  • Molecular Weight : 281.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1803-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1803-00
Availability details for
Quantity
  • Availability
  • Add to Cart