RIE240871082 Rieke Metals, Inc.

3-(4-bromophenyl)-4'-methylpropiophenone

  • MDL number MFCD03843352
  • Molecular Weight : 303.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-bromophenyl)-1-(4-methylphenyl)-1-propanone
Inchi Code 1S/C16H15BrO/c1-12-2-7-14(8-3-12)16(18)11-6-13-4-9-15(17)10-5-13/h2-5,7-10H,6,11H2,1H3
Inchi Key GGHCDQGKIWONCT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1900-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-04
Availability details for
Quantity
  • Availability
  • Add to Cart