RIE240871085 Rieke Metals, Inc.

3-(4-bromophenyl)-4'-methyoxypropiophenone

  • MDL number MFCD03843355
  • Molecular Weight : 319.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1900-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-07
Availability details for
Quantity
  • Availability
  • Add to Cart