RIE240871089 Rieke Metals, Inc.

3-(4-bromophenyl)-2'-carboethoxypropiophenone

  • MDL number MFCD03843359
  • Molecular Weight : 361.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-[3-(4-bromophenyl)propanoyl]benzoate
Inchi Code 1S/C18H17BrO3/c1-2-22-18(21)16-6-4-3-5-15(16)17(20)12-9-13-7-10-14(19)11-8-13/h3-8,10-11H,2,9,12H2,1H3
Inchi Key UOEVAIMFJWTKKO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1900-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-11
Availability details for
Quantity
  • Availability
  • Add to Cart