RIE240871090 Rieke Metals, Inc.

3-(4-bromophenyl)-3'-carboethoxypropiophenone

  • MDL number MFCD03843360
  • Molecular Weight : 361.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-[3-(4-bromophenyl)propanoyl]benzoate
Inchi Code 1S/C18H17BrO3/c1-2-22-18(21)15-5-3-4-14(12-15)17(20)11-8-13-6-9-16(19)10-7-13/h3-7,9-10,12H,2,8,11H2,1H3
Inchi Key NQKFOPVKSJBNFE-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1900-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-12
Availability details for
Quantity
  • Availability
  • Add to Cart