RIE240871094 Rieke Metals, Inc.

3'-bromo-3-(4-bromophenyl)propiophenone

  • MDL number MFCD03843364
  • Molecular Weight : 368.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1900-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-18
Availability details for
Quantity
  • Availability
  • Add to Cart