RIE240871100 Rieke Metals, Inc.

3-(4-bromophenyl)-2',4'-dimethylpropiophenone

  • MDL number MFCD03843371
  • Molecular Weight : 317.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-bromophenyl)-1-(2,4-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H17BrO/c1-12-3-9-16(13(2)11-12)17(19)10-6-14-4-7-15(18)8-5-14/h3-5,7-9,11H,6,10H2,1-2H3
Inchi Key JDXVYABRLIAEQV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1900-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-27
Availability details for
Quantity
  • Availability
  • Add to Cart