RIE240871105 Rieke Metals, Inc.

4'-bromo-3-(4-bromophenyl)-3'-fluoropropiophenone

  • MDL number MFCD03843376
  • Molecular Weight : 386.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromo-3-fluorophenyl)-3-(4-bromophenyl)-1-propanone
Inchi Code 1S/C15H11Br2FO/c16-12-5-1-10(2-6-12)3-8-15(19)11-4-7-13(17)14(18)9-11/h1-2,4-7,9H,3,8H2
Inchi Key KOQBRZUBFGUJQS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1900-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-32
Availability details for
Quantity
  • Availability
  • Add to Cart