RIE240871106 Rieke Metals, Inc.

3-(4-bromophenyl)-4'-chloro-3'-fluoropropiophenone

  • MDL number MFCD03843377
  • Molecular Weight : 341.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1900-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-33
Availability details for
Quantity
  • Availability
  • Add to Cart