RIE240871109 Rieke Metals, Inc.

3-(4-bromophenyl)-2'-fluoropropiophenone

  • MDL number MFCD03843380
  • Molecular Weight : 307.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1900-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-37
Availability details for
Quantity
  • Availability
  • Add to Cart