RIE240871114 Rieke Metals, Inc.

3-(4-bromophenyl)-2'-chloro-4'-fluoropropiophenone

  • MDL number MFCD03843385
  • Molecular Weight : 341.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-bromophenyl)-1-(2-chloro-4-fluorophenyl)-1-propanone
Inchi Code 1S/C15H11BrClFO/c16-11-4-1-10(2-5-11)3-8-15(19)13-7-6-12(18)9-14(13)17/h1-2,4-7,9H,3,8H2
Inchi Key IBZNLSYMKGHYLU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1900-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-50
Availability details for
Quantity
  • Availability
  • Add to Cart