RIE240871116 Rieke Metals, Inc.

3-(4-bromophenyl)-4'-chloro-2'-fluoropropiophenone

  • MDL number MFCD03843387
  • Molecular Weight : 341.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1900-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-52
Availability details for
Quantity
  • Availability
  • Add to Cart