RIE240871117 Rieke Metals, Inc.

3-(4-bromophenyl)-2',3'-dichloropropiophenone

  • MDL number MFCD03843388
  • Molecular Weight : 358.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1900-53

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-53
Availability details for
Quantity
  • Availability
  • Add to Cart