RIE240871121 Rieke Metals, Inc.

3-(4-bromophenyl)-3',5'-dichloropropiophenone

  • MDL number MFCD03843392
  • Molecular Weight : 358.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-bromophenyl)-1-(3,5-dichlorophenyl)-1-propanone
Inchi Code 1S/C15H11BrCl2O/c16-12-4-1-10(2-5-12)3-6-15(19)11-7-13(17)9-14(18)8-11/h1-2,4-5,7-9H,3,6H2
Inchi Key UQSMUDBQSKWBSN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1900-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-57
Availability details for
Quantity
  • Availability
  • Add to Cart