RIE240871124 Rieke Metals, Inc.

3-(4-bromophenyl)-3',5'-difluoropropiophenone

  • MDL number MFCD03843395
  • Molecular Weight : 325.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1900-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-60
Availability details for
Quantity
  • Availability
  • Add to Cart