RIE240871125 Rieke Metals, Inc.

3-(4-bromophenyl)-3',4',5'-trifluoropropiophenone

  • MDL number MFCD03843396
  • Molecular Weight : 343.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-bromophenyl)-1-(3,4,5-trifluorophenyl)-1-propanone
Inchi Code 1S/C15H10BrF3O/c16-11-4-1-9(2-5-11)3-6-14(20)10-7-12(17)15(19)13(18)8-10/h1-2,4-5,7-8H,3,6H2
Inchi Key QYKQGXCTBKERGO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1900-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-61
Availability details for
Quantity
  • Availability
  • Add to Cart