RIE240871126 Rieke Metals, Inc.

3-(4-Bromophenyl)-2',6'-dichloropropiophenone

  • MDL number MFCD08064137
  • Molecular Weight : 358.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1900-62

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-62
Availability details for
Quantity
  • Availability
  • Add to Cart