RIE240871127 Rieke Metals, Inc.

2-(4-bromophenyl)ethyl cyclopropyl ketone

  • MDL number MFCD03843397
  • Molecular Weight : 253.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1901-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1901-00
Availability details for
Quantity
  • Availability
  • Add to Cart