RIE240871131 Rieke Metals, Inc.

3-(4-Bromophenyl)propiophenone

  • MDL number MFCD03843349
  • Molecular Weight : 289.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-bromophenyl)-1-phenyl-1-propanone
Inchi Code 1S/C15H13BrO/c16-14-9-6-12(7-10-14)8-11-15(17)13-4-2-1-3-5-13/h1-7,9-10H,8,11H2
Inchi Key MHNUDVYKHUVNCC-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1900-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-01
Availability details for
Quantity
  • Availability
  • Add to Cart