RIE240871135 Rieke Metals, Inc.

5-Benzoyl-2-chloropyridine

  • MDL number MFCD01754796
  • Molecular Weight : 217.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5625-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5625-01
Availability details for
Quantity
  • Availability
  • Add to Cart