RIE240871139 Rieke Metals, Inc.

4'-bromo-3-(4-bromophenyl)propiophenone

  • MDL number MFCD03843365
  • Molecular Weight : 368.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1900-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1900-19
Availability details for
Quantity
  • Availability
  • Add to Cart