RIE310136458 Rieke Metals, Inc.

2-Chloro-3-pyridylzinc bromide

  • MDL number MFCD18433427
  • Cas Number : 1227833-83-1
  • Molecular Weight : 257.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2303

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2303
Availability details for
Quantity
  • Availability
  • Add to Cart