RIE310136460 Rieke Metals, Inc.

6-Chloro-2-pyridylzinc bromide

  • MDL number MFCD18433429
  • Molecular Weight : 257.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2305

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2305
Availability details for
Quantity
  • Availability
  • Add to Cart