RIE310136461 Rieke Metals, Inc.

3-(1,3-Dioxolan-2-yl)phenylmagnesium bromide

  • MDL number MFCD02260187
  • Molecular Weight : 253.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3089

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3089
Availability details for
Quantity
  • Availability
  • Add to Cart