RIE310136463 Rieke Metals, Inc.

4-(1,3-Dioxolan-2-yl)phenylmagnesium bromide

  • MDL number MFCD02260188
  • Cas Number : 328000-17-5
  • Molecular Weight : 253.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3090

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3090
Availability details for
Quantity
  • Availability
  • Add to Cart