RIE321609117 Rieke Metals, Inc.

Cyclohexyl 4-(2-Methyl-1,3-dioxolan-2-yl)phenyl ketone

  • MDL number MFCD19687271
  • Molecular Weight : 274.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6129-03-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6129-03-01
Availability details for
Quantity
  • Availability
  • Add to Cart