RIE321609132 Rieke Metals, Inc.

1-(4-Chloro-phenyl)-2-(1,3-dioxolan-2-yl)-ethanone

  • MDL number MFCD18433444
  • Molecular Weight : 226.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9016

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9016
Availability details for
Quantity
  • Availability
  • Add to Cart