RIE321609146 Rieke Metals, Inc.

2-(1,3-Dioxolan-2-yl)-1-(4-ethyl-phenyl)-ethanone

  • MDL number MFCD18433458
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9030

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9030
Availability details for
Quantity
  • Availability
  • Add to Cart