RIE321609156 Rieke Metals, Inc.

1-(3,4-Difluoro-phenyl)-2-(1,3-dioxolan-2-yl)-ethanone

  • MDL number MFCD18433468
  • Molecular Weight : 228.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9040

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9040
Availability details for
Quantity
  • Availability
  • Add to Cart