RIE321609177 Rieke Metals, Inc.

2-(1,3-Dioxolan-2-yl)-1-thiophen-3-yl-ethanone

  • MDL number MFCD18433490
  • Molecular Weight : 198.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9062

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9062
Availability details for
Quantity
  • Availability
  • Add to Cart