RIE321609180 Rieke Metals, Inc.

1-(5-bromo-pyridin-3-yl)-2-(1,3-dioxolan-2-yl)-ethanone

  • MDL number MFCD18433493
  • Molecular Weight : 272.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9065

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9065
Availability details for
Quantity
  • Availability
  • Add to Cart