RIE321609181 Rieke Metals, Inc.

1-(1,3-Dioxolan-2-yl)-pentan-2-one

  • MDL number MFCD18433494
  • Molecular Weight : 158.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/9500

Safety Information

MSDS http://riekemetals.com/products-services/catnum/9500
Availability details for
Quantity
  • Availability
  • Add to Cart